Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100917 Supplier Invitrogen™ Supplier No. PA5100917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SNRK.

SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SNRK
Gene Accession No. Q63553, Q8VDU5, Q9NRH2
Gene Alias 2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312
Gene Symbols SNRK
Host Species Rabbit
Immunogen A synthesized peptide derived from human SNRK(Accession Q9NRH2), corresponding to amino acid residues K513-S563.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170837, 20623, 54861
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.