Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRNP200 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107024
Description
Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
Pre-mRNA splicing is catalyzed by the spliceosome, a complex of specialized RNA and protein subunits that removes introns from a transcribed pre-mRNA segment. The spliceosome consists of small nuclear RNA proteins (snRNPs) U1, U2, U4, U5 and U6, together with approximately 80 conserved proteins. U5 snRNP contains nine specific proteins. This gene encodes one of the U5 snRNP-specific proteins. This protein belongs to the DEXH-box family of putative RNA helicases. It is a core component of U4/U6-U5 snRNPs and appears to catalyze an ATP-dependent unwinding of U4/U6 RNA duplices. Mutations in this gene cause autosomal-dominant retinitis pigmentosa. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.
Specifications
SNRNP200 | |
Polyclonal | |
Unconjugated | |
SNRNP200 | |
A330064G03Rik; Activating signal cointegrator 1 complex subunit 3-like 1; ASCC3L1; bad response to refrigeration 2 homolog; BC011390; BRR2; BRR2 homolog; HELIC2; Kiaa0788; RP33; small nuclear ribonucleoprotein 200 (U5); small nuclear ribonucleoprotein 200kDa (U5); small nuclear ribonucleoprotein U5 subunit 200; small nuclear ribonucleoprotein, U5 200kDa subunit; SNRNP200; U5 small nuclear ribonucleoprotein 200 kDa helicase; U5 snRNP-specific 200 kDa protein; U5 snRNP-specific protein, 200 kDa (DEXH RNA helicase family); U5-200KD; U5-200-KD | |
Rabbit | |
Affinity chromatography | |
RUO | |
23020 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
O75643 | |
SNRNP200 | |
Synthetic peptide located within the following region: GDEDVYGEVR EEASDDDMEG DEAVVRCTLS ANLVASGELM SSKKKDLHPR. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction