Learn More
SNRNP200 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Specifications
Specifications
| Antigen | SNRNP200 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | SNRNP200 |
| Gene Accession No. | O75643, Q6P4T2 |
| Gene Alias | activating signal cointegrator 1 complex subunit 3-like 1, ASCC3L1, bad response to refrigeration 2 homolog, BRR2, BRR2 homolog, HELIC2, KIAA0788, retinitis pigmentosa 33 (autosomal domit), RP33, small nuclear ribonucleoprotein 200kDa (U5), small nuclear ribonucleoprotein, U5 200kDa subunit, U5 snRNP specific protein, 200 KD, U5 snRNP-specific 200 kDa protein, U5-200KD |
| Gene Symbols | SNRNP200 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.