Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRNP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559473 Supplier Invitrogen™ Supplier No. PA559473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMPVVVVQSA TVLDLKKAIQ RYVQLKQERE GGIQHISWSY VWRTYHLTSA GEKLTEDRKK LRDYGIRNRD E Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP25
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNRNP25
Gene Accession No. Q9BV90
Gene Alias C16orf33; minus-99 protein; small nuclear ribonucleoprotein 25kDa (U11/U12); small nuclear ribonucleoprotein U11/U12 subunit 25; small nuclear ribonucleoprotein, U11/U12 25kDa subunit; SNRNP25; U11/U12 small nuclear ribonucleoprotein 25 kDa protein; U11/U12 snRNP 25 kDa protein; U11/U12 snRNP 25K
Gene Symbols SNRNP25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNRNP25. Recombinant protein control fragment (Product #RP-98708).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.