Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRNP27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556970
Description
Immunogen sequence: TSPSPSRLKE RRDEEKKETK ETKSKERQIT EEDLEGKTEE EIEMMKLMGF ASFDSTKGKK VDGSVNAYAI NVSQKRKYRQ YMNRKGGFNR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
May play a role in mRNA splicing.
Specifications
SNRNP27 | |
Polyclonal | |
Unconjugated | |
SNRNP27 | |
2610209M04Rik; 27K; AI449063; AU022368; Nucleic acid-binding protein RY-1; putative nucleic acid binding protein RY-1; RY1; small nuclear ribonucleoprotein 27 (U4/U6.U5); small nuclear ribonucleoprotein 27kDa (U4/U6.U5); small nuclear ribonucleoprotein U4/U6.U5 subunit 27; small nuclear ribonucleoprotein, U4/U6.U5 27kDa subunit; Snrnp27; U4/U6.U5 small nuclear ribonucleoprotein 27 kDa protein; U4/U6.U5 snRNP 27 kDa protein; U4/U6.U5 tri-snRNP-associated 27 kDa protein; U4/U6.U5 tri-snRNP-associated protein 3; U4/U6.U5-27K; U4/U6U5-27K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVK2 | |
SNRNP27 | |
Recombinant protein corresponding to Human SNRNP27. Recombinant protein control fragment (Product #RP-96065). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction