Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRNP35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541831 Supplier Invitrogen™ Supplier No. PA541831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RAVWRAMLAR YVPNKGVIGD PLLTLFVARL NLQTKEDKLK EVFSRYGDIR Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.

U1SNRNPBP is a homolog of U1-snRNP binding protein. Its N-terminal half contains a RNA recognition motif and the C-terminal half is rich in Arg/Asp and Arg/Glu dipeptides, a characteristic of a variety of splicing factors.The protein encoded by this gene is a homolog of U1-snRNP binding protein. Its N-terminal half contains a RNA recognition motif and the C-terminal half is rich in Arg/Asp and Arg/Glu dipeptides, a characteristic of a variety of splicing factors. This gene is differentially expressed in a variety of human tissues. Multiple alternatively spliced transcript variants have been found for this gene, and they differ in the 5' sequence regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP35
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SNRNP35
Gene Accession No. Q16560
Gene Alias 6330548G22Rik; HM1; HM-1; Protein HM-1; RGD1310724; small nuclear ribonucleoprotein 35 (U11/U12); small nuclear ribonucleoprotein 35kDa (U11/U12); small nuclear ribonucleoprotein U11/U12 subunit 35; small nuclear ribonucleoprotein, U11/U12 35kDa subunit; SNRNP35; U1 snRNP binding protein homolog; U1 snRNP-binding protein homolog; U11/U12 small nuclear ribonucleoprotein 35 kDa protein; U11/U12 snRNP 35 kDa protein; U11/U12 snRNP 35K; U11/U12-35K; U1-snRNP binding protein homolog; U1snrnpbp
Gene Symbols SNRNP35
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human U1SNRNPBP (aa 29-78).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11066
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.