Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRNP35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531496 Supplier Invitrogen™ Supplier No. PA531496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, mouse brain. Predicted reactivity: Mouse (88%), Rat (87%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

U1SNRNPBP is a homolog of U1-snRNP binding protein. Its N-terminal half contains a RNA recognition motif and the C-terminal half is rich in Arg/Asp and Arg/Glu dipeptides, a characteristic of a variety of splicing factors.The protein encoded by this gene is a homolog of U1-snRNP binding protein. Its N-terminal half contains a RNA recognition motif and the C-terminal half is rich in Arg/Asp and Arg/Glu dipeptides, a characteristic of a variety of splicing factors. This gene is differentially expressed in a variety of human tissues. Multiple alternatively spliced transcript variants have been found for this gene, and they differ in the 5' sequence regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SNRNP35
Gene Accession No. Q16560, Q9D384
Gene Alias 6330548G22Rik; HM1; HM-1; Protein HM-1; RGD1310724; small nuclear ribonucleoprotein 35 (U11/U12); small nuclear ribonucleoprotein 35kDa (U11/U12); small nuclear ribonucleoprotein U11/U12 subunit 35; small nuclear ribonucleoprotein, U11/U12 35kDa subunit; SNRNP35; U1 snRNP binding protein homolog; U1 snRNP-binding protein homolog; U11/U12 small nuclear ribonucleoprotein 35 kDa protein; U11/U12 snRNP 35 kDa protein; U11/U12 snRNP 35K; U11/U12-35K; U1-snRNP binding protein homolog; U1snrnpbp
Gene Symbols SNRNP35
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human U1SNRNPBP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11066, 76167
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.