Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRNP40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555331
Description
Immunogen sequence: QGNVHNFEKN LLRCSWSPDG SKIAAGSADR FVYVWDTTSR RILYKLPGHA GSINEVAFHP DEPIIISASS DKRLYMGEIQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
This gene encodes a component of the U5 small nuclear ribonucleoprotein (snRNP) particle. The U5 snRNP is part of the spliceosome, a multiprotein complex that catalyzes the removal of introns from pre-messenger RNAs.
Specifications
SNRNP40 | |
Polyclonal | |
Unconjugated | |
SNRNP40 | |
0610009C03Rik; 38 kDa-splicing factor; 40K; HPRP8BP; Prp8-binding protein; Prp8bp; PRPF8BP; RGD1309198; RP11-490K7.3; SFP38; small nuclear ribonucleoprotein 40 (U5); small nuclear ribonucleoprotein 40kDa (U5); small nuclear ribonucleoprotein U5 subunit 40; small nuclear ribonucleoprotein, U5 40kDa subunit; Snrnp40; SPF38; U5 small nuclear ribonucleoprotein 40 kDa protein; U5 snRNP 40 kDa protein; U5 snRNP-specific 40 kDa protein; U5 snRNP-specific 40 kDa protein (hPrp8-binding); U5 snRNP-specific protein (Prp8-binding); U5-40K; U5-40kD protein; WD repeat domain 57 (U5 snRNP specific); WD repeat-containing protein 57; Wdr57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
313056, 66585, 9410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PE01, Q96DI7 | |
SNRNP40 | |
Recombinant protein corresponding to Human SNRNP40. Recombinant protein control fragment (Product #RP-94001). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction