Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRNP40 Polyclonal Antibody
SNRNP40 Polyclonal Antibody
Supplier: Thermo Scientific PA555589
Description
SNRNP40 Polyclonal Antibody for Western Blot, IHC

Specifications
SNRNP40 | |
Polyclonal | |
Unconjugated | |
SNRNP40 | |
0610009C03Rik; 38 kDa-splicing factor; 40K; HPRP8BP; Prp8-binding protein; Prp8bp; PRPF8BP; RGD1309198; RP11-490K7.3; SFP38; small nuclear ribonucleoprotein 40 (U5); small nuclear ribonucleoprotein 40kDa (U5); small nuclear ribonucleoprotein U5 subunit 40; small nuclear ribonucleoprotein, U5 40kDa subunit; Snrnp40; SPF38; U5 small nuclear ribonucleoprotein 40 kDa protein; U5 snRNP 40 kDa protein; U5 snRNP-specific 40 kDa protein; U5 snRNP-specific 40 kDa protein (hPrp8-binding); U5 snRNP-specific protein (Prp8-binding); U5-40K; U5-40kD protein; WD repeat domain 57 (U5 snRNP specific); WD repeat-containing protein 57; Wdr57 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DI7 | |
SNRNP40 | |
Recombinant protein corresponding to Human SNRNP40. Recombinant protein control fragment (Product #RP-94000). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction