Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRNP48 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186760
Description
SNRNP48 Polyclonal specifically detects SNRNP48 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SNRNP48 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
C6orf151, chromosome 6 open reading frame 151, dJ336K20B.1, dJ512B11.2, FLJ32234, MGC138904, MGC138905, small nuclear ribonucleoprotein 48kDa (U11/U12), U11/U12 small nuclear ribonucleoprotein 48 kDa protein, U11/U12 snRNP 48 kDa protein, U11/U12 snRNP 48K, U11/U12-48K | |
Rabbit | |
Affinity Purified | |
RUO | |
154007 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SNRNP48 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QEELNEFVESGCRTLEEVTASLGWDLDSLDPGEEEAAEDEVVICPYDSNHHMPKSSLAKHMASCRLRKMGYTKEEEDEMYNPEFFYENVKIP | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction