Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRNP70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559843 Supplier Invitrogen™ Supplier No. PA559843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDPIPYLPPL EKLPHEKHHN QPYCGIAPYI REFEDPRDAP PPTRAETREE RMERKRREKI ERRQQEVETE LKMWDPHNDP NAQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

SNRP70 contains 1 RRM (RNA recognition motif) domain and mediates the splicing of pre-mRNA by binding to the loop I region of U1-snRNA. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP70
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNRNP70
Gene Accession No. P08621
Gene Alias 2700022N21Rik; 3200002N22Rik; AI325098; LOW QUALITY PROTEIN: U1 small nuclear ribonucleoprotein 70 kDa; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; small nuclear ribonucleoprotein 70 (U1); small nuclear ribonucleoprotein 70kDa; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen); small nuclear ribonucleoprotein U1 subunit 70; small nuclear ribonucleoprotein, U1 70kDa subunit; Snp1; Snrnp70; SNRP70; snrp70 protein; Srnp70; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 small ribonucleoprotein 70kd; U1 snRNP 70 kDa; U1-70; U170K; U1-70K; U1AP; U1AP1; U1RNP; Unknown (protein for MGC:138058); wu:fc20b04; zgc:136450
Gene Symbols SNRNP70
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNRNP70. Recombinant protein control fragment (Product #RP-102919).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.