Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRNP70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541728 Supplier Invitrogen™ Supplier No. PA541728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PHNDPNAQGD AFKTLFVARV NYDTTESKLR REFEVYGPIK RIHMVYSKRS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

SNRP70 contains 1 RRM (RNA recognition motif) domain and mediates the splicing of pre-mRNA by binding to the loop I region of U1-snRNA. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP70
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SNRNP70
Gene Accession No. P08621
Gene Alias 2700022N21Rik; 3200002N22Rik; AI325098; LOW QUALITY PROTEIN: U1 small nuclear ribonucleoprotein 70 kDa; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; small nuclear ribonucleoprotein 70 (U1); small nuclear ribonucleoprotein 70kDa; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen); small nuclear ribonucleoprotein U1 subunit 70; small nuclear ribonucleoprotein, U1 70kDa subunit; Snp1; Snrnp70; SNRP70; snrp70 protein; Srnp70; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 small ribonucleoprotein 70kd; U1 snRNP 70 kDa; U1-70; U170K; U1-70K; U1AP; U1AP1; U1RNP; Unknown (protein for MGC:138058); wu:fc20b04; zgc:136450
Gene Symbols SNRNP70
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SNRP70 (aa 91-140).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6625
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.