Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561243
Description
Immunogen sequence: IIAKMKGTFV ERDRKREKRK PKSQETPATK KAVQGGGATP VVGAVQGPVP GMPPMTQAPR IMHHMPGQPP YMP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.
The protein encoded by this gene associates with stem loop II of the U1 small nuclear ribonucleoprotein, which binds the 5' splice site of precursor mRNAs and is required for splicing. The encoded protein autoregulates itself by polyadenylation inhibition of its own pre-mRNA via dimerization and has been implicated in the coupling of splicing and polyadenylation.
Specifications
SNRPA | |
Polyclonal | |
Unconjugated | |
SNRPA | |
C430021M15Rik; fc19d01; MGC137110 protein; Mud1; nRNP A; nuclear ribonucleoprotein A; protein A of the U1 small nuclear RNA; Rnu1a1; Rnu1a-1; small nuclear ribonucleoprotein polypeptide A; Snrpa; U1 small nuclear ribonucleoprotein A; U1 small nuclear RNP-specific A; U1 snRNP A; U1 snRNP protein A; U1 snRNP-specific protein A; U1A; U1-A; Unknown (protein for MGC:77810); wu:fc19d01; zgc:77810 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6626 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09012 | |
SNRPA | |
Recombinant protein corresponding to Human SNRPA. Recombinant protein control fragment (Product #RP-98877). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction