Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPA Recombinant Rabbit Monoclonal Antibody (JB33-48)
GREENER_CHOICE

Catalog No. PIMA534722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534722 Supplier Invitrogen™ Supplier No. MA534722
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat brain tissue lysate, K562, A431, mouse kidney tissue lysate, 293T, SH-SY-5Y, SiHa human lung cancer tissue, mouse liver tissue.

The protein encoded by this gene associates with stem loop II of the U1 small nuclear ribonucleoprotein, which binds the 5' splice site of precursor mRNAs and is required for splicing. The encoded protein autoregulates itself by polyadenylation inhibition of its own pre-mRNA via dimerization and has been implicated in the coupling of splicing and polyadenylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPA
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB33-48
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SNRPA
Gene Accession No. P09012
Gene Alias C430021M15Rik; fc19d01; MGC137110 protein; Mud1; nRNP A; nuclear ribonucleoprotein A; protein A of the U1 small nuclear RNA; Rnu1a1; Rnu1a-1; small nuclear ribonucleoprotein polypeptide A; Snrpa; U1 small nuclear ribonucleoprotein A; U1 small nuclear RNP-specific A; U1 snRNP A; U1 snRNP protein A; U1 snRNP-specific protein A; U1A; U1-A; Unknown (protein for MGC:77810); wu:fc19d01; zgc:77810
Gene Symbols SNRPA
Host Species Rabbit
Immunogen Recombinant protein within Human U1A aa 1-96.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6626
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.