Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPA1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501566496
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6496 Supplier Bethyl Laboratories, Inc Supplier No. A303948A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Principal Names: Small nuclear ribonucleoprotein polypeptide A; SNRPA1; U2 snRNP-A; Official Gene Symbol: SNRPA1 Gene ID: 6627 (Human) Gene Map Locus: 15q26.3 (Human) SNRPA1, a 28KDa protein, isa specific component of U2 snRNP particle andconsists of a unique N-terminal leucine-rich region and an extremely hydrophobic RNA-binding C-terminal region. Itlacks segments homologous to RNP1 and RNP2, commonA.A motifs found in several RNA-binding proteins. It associates with U2 snRNP and participates in transcription and pre-mRNA splicing. It might play an important role in cell division and proliferation. Patients with connective tissue disorders often produce autoantibodies against snRNP particles.

Specifications

Antigen SNRPA1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SNRPA1
Gene Accession No. P09661, P57784
Gene Alias U2 small nuclear ribonucleoprotein A′; U2 snRNP-A′; SNRPA1
Gene Symbols Snrpa1
Host Species Rabbit
Immunogen Between 205 and 255
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6627, 68981
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.