Learn More
Invitrogen™ SNRPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527559
Description
Recommended positive controls: HeLa nucleus. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (93%), Xenopus laevis (89%), Chicken (96%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.
Specifications
| SNRPB | |
| Polyclonal | |
| Unconjugated | |
| SNRPB | |
| AL024368; AU018828; B polypeptide of Sm protein; CCMS; COD; sm protein B; Sm protein B/B'; SM11; small nuclear ribonucleoprotein B; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; small nuclear ribonucleoprotein polypeptides B and B1; small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; sm-B; Sm-B/B'; SmB/B'; sm-B/Sm-B'; SmB/SmB'; SNRNP-B; Snrpb; SNRPB1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, RNA Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.38 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P14678 | |
| SNRPB | |
| Recombinant protein encompassing a sequence within the center region of human SNRPB. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.