Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110407 Supplier Invitrogen™ Supplier No. PA5110407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTVGKSSKML QHIDYRMRCI LQDGRIFIGT FKAFDKHMNL ILCDCDEFRK IKPKNSKQAE REEKRVLGLV LLRGENLVSM TVEGPPPKDT GIARVPLAGA.

The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNRPB
Gene Accession No. P14678, P27048
Gene Alias AL024368; AU018828; B polypeptide of Sm protein; CCMS; COD; sm protein B; Sm protein B/B'; SM11; small nuclear ribonucleoprotein B; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; small nuclear ribonucleoprotein polypeptides B and B1; small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; sm-B; Sm-B/B'; SmB/B'; sm-B/Sm-B'; SmB/SmB'; SNRNP-B; Snrpb; SNRPB1
Gene Symbols SNRPB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SNRPB (NP_003082.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171365, 20638, 6628
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.