Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRPB2 Antibody (OTI2A5), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274260UV
Description
SNRPB2 Monoclonal specifically detects SNRPB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SNRPB2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SNRPB2 | |
Full length recombinant protein of human SNRPB2 (NP_937863) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2A5 | |
Western Blot, Immunohistochemistry | |
MGC24807, MGC45309, Msl1, small nuclear ribonucleoprotein polypeptide B, small nuclear ribonucleoprotein polypeptide B′′, small nuclear ribonucleoprotein polypeptide B2, U2 small nuclear ribonucleoprotein B′′, U2 snRNP B′′ | |
Mouse | |
Affinity Purified | |
Immunology | |
6629.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction