Learn More
Invitrogen™ SNRPB2 Monoclonal Antibody (2F4)

Description
Peptide Sequence: MDIRPNHTIY INNMNDKIKK EELKRSLYAL FSQFGHVVDI VALKTMKMRG QAFVIFKELG SSTNALRQLQ GFPFYGKPMR IQYAKTDSDI ISKMRGTFAD KEKKKEKKKA KTVEQTATTT NKKPGQGTPN SANTQGNSTP NPQVPDYPPN YILFLNNLPE ETNEMMLSML FNQFPGFKEV RLVPGRHDIA FVEFENDGQA GAARDALQGF KITPSHAMKI TYAKK.
Specifications
Specifications
| Antigen | SNRPB2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2F4 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SNRPB2 |
| Gene Accession No. | P08579 |
| Gene Alias | 2810052G09Rik; Msl1; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptide B''; small nuclear ribonucleoprotein polypeptide B2; Snrpb2; U2 small nuclear ribonucleoprotein B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''; U2B'' |
| Gene Symbols | SNRPB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.