Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPB2 Monoclonal Antibody (OTI2A5)
GREENER_CHOICE

Catalog No. PIMA526940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526940 Supplier Invitrogen™ Supplier No. MA526940
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNRPB2 Monoclonal Antibody for Western Blot, IHC (P)

SNRPB2 is involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A' protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SNRPB2
Gene Accession No. P08579
Gene Alias 2810052G09Rik; Msl1; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptide B''; small nuclear ribonucleoprotein polypeptide B2; Snrpb2; U2 small nuclear ribonucleoprotein B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''; U2B''
Gene Symbols SNRPB2
Host Species Mouse
Immunogen Full length human recombinant protein of SNRPB2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6629
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.