Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRPB2 Monoclonal Antibody (OTI2E8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA808141

Description
Involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A′ protein.Specifications
SNRPB2 | |
Monoclonal | |
Unconjugated | |
SNRPB2 | |
Msl1, U2B′′ | |
Mouse | |
Affinity Chromatography | |
RUO | |
6629 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2E8 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P08579 | |
SNRPB2 | |
Full length human recombit protein of human SNRPB2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction