Learn More
SNRPB2 Mouse anti-Human, Clone: OTI1B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808091

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A′ protein.Specifications
SNRPB2 | |
Monoclonal | |
Unconjugated | |
SNRPB2 | |
Msl1, U2B′′ | |
Mouse | |
Affinity Chromatography | |
RUO | |
6629 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08579 | |
SNRPB2 | |
Full length human recombit protein of human SNRPB2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.