Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584991
Description
Immunogen sequence: AKTDSDIISK MRGTFADKEK KKEKKKAKTV EQTATTTNKK PGQGTPNSAN TQ.
SNRPB2 is involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A' protein.
Specifications
SNRPB2 | |
Polyclonal | |
Unconjugated | |
SNRPB2 | |
2810052G09Rik; Msl1; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptide B''; small nuclear ribonucleoprotein polypeptide B2; Snrpb2; U2 small nuclear ribonucleoprotein B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''; U2B'' | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P08579 | |
SNRPB2 | |
Recombinant protein corresponding to Human SNRPB2. Recombinant protein control fragment (Product #RP-108660). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction