Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRPD1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306017F
Description
SNRPD1 Polyclonal antibody specifically detects SNRPD1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SNRPD1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
HsT2456, small nuclear ribonucleoprotein D1 polypeptide 16kDa, small nuclear ribonucleoprotein Sm D1, Sm-D autoantigen, Sm-D1, SMD1, snRNP core protein D1, SNRPD | |
Produced in rabbits immunized with E. coli-derived Human SNRPD1 fragment. | |
0.1 mL | |
Immunology | |
6632 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction