Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512459
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512459 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512459 Supplier Invitrogen™ Supplier No. PA512459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, mouse and non-human primate based on sequence homology.

SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPD1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SNRPD1
Gene Accession No. P62314
Gene Alias AA407109; AL023031; CHUNP6882; fk26a01; HsT2456; small nuclear ribonucleoprotein D1; small nuclear ribonucleoprotein D1 polypeptide; small nuclear ribonucleoprotein D1 polypeptide 16kDa; small nuclear ribonucleoprotein D1 polypeptide 16kDa pseudogene; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; small nuclear ribonucleoprotein Sm D1; Sm-D autoantigen; SMD1; sm-D1; snprd1; snRNP core protein D1; snrnpd1; SNRPD; Snrpd1; snrpd1 protein; wu:fk26a01; zgc:86929
Gene Symbols SNRPD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 69-98 amino acids from the C-terminal region of human SNRPD1.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6632
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.