Learn More
SNRPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 103521AP150UL

Description
This antibody detects 13-16 kDa (SMD1) and 30 kDa (SMD1/SMD2 dimer form).
SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.Specifications
SNRPD1 | |
Polyclonal | |
Unconjugated | |
SNRPD1 | |
HsT2456, Sm D autoantigen, Sm D1, SMD1, snRNP core protein D1, SNRPD, SNRPD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20641, 291794, 6632 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62314, P62315 | |
Snrpd1 | |
SNRPD1 Fusion Protein Ag0364 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.