Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPD1 Recombinant Mouse Monoclonal Antibody (24E2)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550236
Description
SNRPD1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA
SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.
Specifications
SNRPD1 | |
Recombinant Monoclonal | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P62314 | |
SNRPD1 | |
Recombinant Human SNRPD1 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
24E2 | |
Unconjugated | |
SNRPD1 | |
AA407109; AL023031; CHUNP6882; fk26a01; HsT2456; small nuclear ribonucleoprotein D1; small nuclear ribonucleoprotein D1 polypeptide; small nuclear ribonucleoprotein D1 polypeptide 16kDa; small nuclear ribonucleoprotein D1 polypeptide 16kDa pseudogene; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; small nuclear ribonucleoprotein Sm D1; Sm-D autoantigen; SMD1; sm-D1; snprd1; snRNP core protein D1; snrnpd1; SNRPD; Snrpd1; snrpd1 protein; wu:fk26a01; zgc:86929 | |
Mouse | |
Affinity chromatography | |
RUO | |
6632 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction