Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593328 Supplier Invitrogen™ Supplier No. PA593328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLLNKPKSE MTPEELQKRE EEEFNTGPLS VLTQSVKNNT QVLINCRNNK KLLGRVKAFD RHCNMVLENV KEMWTEVPKS GKGKKKSKPV NKDRYISKMF LRGDSVIVVL RNPLIAGK; Positive Samples: MCF7, HL-60, HepG2, HeLa, Jurkat, Rat testis, Rat heart; Cellular Location: Cytoplasm, Nucleus, cytosol.

The protein encoded by this gene belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SNRPD2
Gene Accession No. P62316, P62317
Gene Alias 1810009A06Rik; small nuclear ribonucleoprotein D2; small nuclear ribonucleoprotein D2 polypeptide; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; small nuclear ribonucleoprotein Sm D2; SMD2; sm-D2; snRNP core protein D2; SNRPD1; SNRPD2
Gene Symbols SNRPD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human SNRPD2 (NP_0045881).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107686, 6633, 680309
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.