Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103307 Supplier Invitrogen™ Supplier No. PA5103307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SmD3.

This gene encodes a core component of the spliceosome, which is a nuclear ribonucleoprotein complex that functions in pre-mRNA splicing. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Snrpd3
Gene Accession No. P62318, P62320
Gene Alias 1700043E15Rik; 2310009E13Rik; AW046420; small nuclear ribonucleoprotein D3; small nuclear ribonucleoprotein D3 polypeptide; small nuclear ribonucleoprotein D3 polypeptide 18kDa; small nuclear ribonucleoprotein Sm D3; SMD3; Sm-D3; snRNP core protein D3; SNRPD3
Gene Symbols Snrpd3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SNRPD3(Accession P62318), corresponding to amino acid residues L58-A108.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6634, 67332, 687711
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.