Learn More
Invitrogen™ SNRPD3 Polyclonal Antibody

Description
Positive Control: Jurkat cell lysate, mouse lung tissue lysate, MCF-7 cell lysates, human colorectal carcinoma tissue, human kidney tissue. Subcellular Location: cytosol, Nucleus.
Specifications
Specifications
| Antigen | SNRPD3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Snrpd3 |
| Gene Accession No. | P62318, P62320 |
| Gene Alias | 1700043E15Rik; 2310009E13Rik; AW046420; small nuclear ribonucleoprotein D3; small nuclear ribonucleoprotein D3 polypeptide; small nuclear ribonucleoprotein D3 polypeptide 18kDa; small nuclear ribonucleoprotein Sm D3; SMD3; Sm-D3; snRNP core protein D3; SNRPD3 |
| Gene Symbols | Snrpd3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.