Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570618
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
The protein encoded by SNRPE is a core component of U small nuclear ribonucleoproteins, which are key components of the pre-mRNA processing spliceosome. The encoded protein plays a role in the 3' end processing of histone transcripts. This protein is one of the targets in the autoimmune disease systemic lupus erythematosus, and mutations in SNRPE have been associated with hypotrichosis. Several pseudogenes of SNRPE have been identified.
Specifications
SNRPE | |
Polyclonal | |
Unconjugated | |
SNRPE | |
AL022645; B raf; Braf; B-raf; C76690; HYPT11; sm protein E; small nuclear ribonucleoprotein E; small nuclear ribonucleoprotein polypeptide E; smE; Sm-E; snRNPE; snRNP-E; SNRPE | |
Rabbit | |
Affinity chromatography | |
RUO | |
6635 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P62304 | |
SNRPE | |
synthetic peptide directed towards the middle region of human SNRPE (aa 30-79). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction