Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598165
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598165 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598165 Supplier Invitrogen™ Supplier No. PA598165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNRPE Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The protein encoded by SNRPE is a core component of U small nuclear ribonucleoproteins, which are key components of the pre-mRNA processing spliceosome. The encoded protein plays a role in the 3' end processing of histone transcripts. This protein is one of the targets in the autoimmune disease systemic lupus erythematosus, and mutations in SNRPE have been associated with hypotrichosis. Several pseudogenes of SNRPE have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPE
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SNRPE
Gene Accession No. P62304
Gene Alias AL022645; B raf; Braf; B-raf; C76690; HYPT11; sm protein E; small nuclear ribonucleoprotein E; small nuclear ribonucleoprotein polypeptide E; smE; Sm-E; snRNPE; snRNP-E; SNRPE
Gene Symbols SNRPE
Host Species Rabbit
Immunogen Recombinant Human Small nuclear ribonucleoprotein E protein (1-92AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6635
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.