Learn More
SNRPE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 204071AP150UL

Description
This antibody raises against the full length of human SNRPE, and can recognize two bands of SNRPE, include a 9 kDa band (isoform) and a 11 kDa band (SNRPE).
U1, U2, U5 and U4/U6 are small nuclear ribonucleoproteins (snRNPs) that comprise the spliceosome in eukaryotes. Each UsnRNP contains common Sm proteins B/B′, D1, D2, D3, E, F and G. The Sm proteins pair up as D1-D2, B/B′-D3 and E-F-G to form RNA-free hetero-oligomers in the cytoplasm. Sm proteins aid in the cytoplasmic construction of the UsnRNPs by binding to a conserved Sm site on UsnRNA and forming a stable snRNP core complex.Specifications
SNRPE | |
Polyclonal | |
Unconjugated | |
SNRPE | |
Sm E, Sm protein E, SME, snRNP E, SNRPE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100360682, 20643, 6635 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62304, P62305 | |
SNRPE | |
SNRPE Fusion Protein Ag14227 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.