Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRPF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SNRPF |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SNRPF Polyclonal specifically detects SNRPF in Human samples. It is validated for Western Blot.Specifications
SNRPF | |
Polyclonal | |
Rabbit | |
PBSCF, Sm protein F, small nuclear ribonucleoprotein polypeptide F, SMF, Sm-Fsmall nuclear ribonucleoprotein F, snRNP-F | |
SNRPF | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
6636 | |
Synthetic peptides corresponding to SNRPF(small nuclear ribonucleoprotein polypeptide F) The peptide sequence was selected from the middle region of SNRPF. Peptide sequence GYMNMQLANTEEYIDGALSGHLGEVLIRCNNVLYIRGVEEEEEDGEMRE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title