Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ SNRPF Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541766
Description
Peptide sequence: MSLPLNPKPF LNGLTGKPVM VKLKWGMEYK GYLVSVDGYM NMQLANTEEY Sequence homology: Cow: 100%; Dog: 100%; Goat: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%; Zebrafish: 100%.
SNRPF belongs to the snRNP Sm proteins family and is associated with snRNP U1, U2, U4/U6 and U5. 
            Specifications
| SNRPF | |
| Polyclonal | |
| Unconjugated | |
| SNRPF | |
| 2010013O18Rik; LOC615240 protein; PBSCF; Sm protein F; Small nuclear ribonucleoprotein F; small nuclear ribonucleoprotein polypeptide F; SMF; sm-F; snRNP-F; snrpf; snrpfl | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6636 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62306 | |
| SNRPF | |
| Synthetic peptide directed towards the N-terminal of human SNRPF (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                            