Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588388 Supplier Invitrogen™ Supplier No. PA588388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLPLNPKPF LNGLTGKPVM VKLKWGMEYK GYLVSVDGYM NMQLANTEEY IDGALSGHLG EVLIRCNNVL YIRGVEEEEE DGEMRE; Positive Samples: HeLa, HT-29, 293T, LO2, SGC-7901, Mouse brain, Mouse testis; Cellular Location: Cytoplasm, Nucleus, cytosol.

SNRPF belongs to the snRNP Sm proteins family and is associated with snRNP U1, U2, U4/U6 and U5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPF
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNRPF
Gene Accession No. P62306, P62307
Gene Alias 2010013O18Rik; LOC615240 protein; PBSCF; Sm protein F; Small nuclear ribonucleoprotein F; small nuclear ribonucleoprotein polypeptide F; SMF; sm-F; snRNP-F; snrpf; snrpfl
Gene Symbols SNRPF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-86 of human SNRPF (NP_0030861).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6636, 680737, 69878
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.