Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112620
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112620 Supplier Invitrogen™ Supplier No. PA5112620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNRPG Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Core component of the spliceosomal U1, U2, U4 and U5 small nuclear ribonucleoproteins (snRNPs), the building blocks of the spliceosome. Thereby, plays an important role in the splicing of cellular pre-mRNAs. Most spliceosomal snRNPs contain a common set of Sm proteins SNRPB, SNRPD1, SNRPD2, SNRPD3, SNRPE, SNRPF and SNRPG that assemble in an heptameric protein ring on the Sm site of the small nuclear RNA to form the core snRNP. Appears to function in the U7 snRNP complex that is involved in histone 3'- end processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPG
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SNRPG
Gene Accession No. P62308
Gene Alias 2810024K17Rik; AL022803; PBSCG; Sm protein G; Small nuclear ribonucleoprotein G; small nuclear ribonucleoprotein polypeptide G; SMG; Sm-G; snRNP-G; Snrpg
Gene Symbols SNRPG
Host Species Rabbit
Immunogen Recombinant Human Small nuclear ribonucleoprotein G protein (1-76AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6637
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.