Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115944
Description
Antibody detects endogenous levels of total SNRPN.
The protein encoded by this gene is one polypeptide of a small nuclear ribonucleoprotein complex and belongs to the snRNP SMB/SMN family. The protein plays a role in pre-mRNA processing, possibly tissue-specific alternative splicing events. Although individual snRNPs are believed to recognize specific nucleic acid sequences through RNA-RNA base pairing, the specific role of this family member is unknown. The protein arises from a bicistronic transcript that also encodes a protein identified as the SNRPN upstream reading frame (SNURF). Multiple transcription initiation sites have been identified and extensive alternative splicing occurs in the 5' untranslated region. Additional splice variants have been described but sequences for the complete transcripts have not been determined. The 5' UTR of this gene has been identified as an imprinting center. Alternative splicing or deletion caused by a translocation event in this paternally-expressed region is responsible for Angelman syndrome or Prader-Willi syndrome due to parental imprint switch failure.
Specifications
| SNRPN | |
| Polyclonal | |
| Unconjugated | |
| SNRPN | |
| 2410045I01Rik; DKFZp686C0927; DKFZp686M12165; DKFZp761I1912; DKFZp762N022; FE 294; FE 294 psi; FE294; FE294 gene for snRNP-associated polypeptide N; FE294 psi; FE294 psi pseudogene; FLJ33569; FLJ36996; FLJ39265; HCERN3; MGC29886; Peg4; PWCR; Pwcr1; RT-LI; Sm protein D; sm protein N; small nuclear ribonucleoparticle-associated protein (snRNP) mRNA clone Sm51; small nuclear ribonucleoparticle-associated protein (snRNP) mRNA, clone Sm51; small nuclear ribonucleoprotein N; small nuclear ribonucleoprotein polypeptide N; small nuclear ribonucleoprotein-associated protein N; Sm-D; Smn; sm-N; snRNP-N; Snrpn; SNRPN upstream reading frame; SNURF; SNURF-SNRPN; tissue-specific splicing protein; tissue-specific-splicing protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20646, 6638 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P63162, P63163 | |
| SNRPN | |
| A synthesized peptide derived from human SNRPN(Accession P63162), corresponding to amino acid residues A157-P207. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction