Learn More
SNTA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305352AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Syntrophin alpha-1 (Dystrophin, SNTA1) is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.Specifications
SNTA1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q13424 | |
SNTA1 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SNTA1 | |
RP23-192L3.1, AW228934, Snt1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6640 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.