Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNTB1 Polyclonal Antibody

Catalog No. PIPA5121592
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121592 20 μL
PIPA5119452 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121592 Supplier Invitrogen™ Supplier No. PA5121592
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNTB1 Polyclonal Antibody for Western Blot, IHC (P)

Adapter protein that binds to and probably organizes the subcellular localization of a variety of membrane proteins. May link various receptors to the actin cytoskeleton and the dystrophin glycoprotein complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNTB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SNTB1
Gene Accession No. Q13884, Q99L88
Gene Alias 59 kDa dystrophin-associated protein A1 basic component 1; 59-1 DAP; 59-DAP; A1B; beta-1-syntrophin; beta1-Syntrophin; BSYN2; DAPA1B; dystrophin-associated protein A1, 59kD, basic component 1; SNT2; SNT2B1; SNTB1; syntrophin beta 1; syntrophin, basic 1; syntrophin, beta 1; syntrophin, beta 1 (dystrophin-associated protein A1, 59kDa, basic component 1); syntrophin-2; Tax interaction protein 43; TIP-43
Gene Symbols SNTB1
Host Species Rabbit
Immunogen Recombinant protein Syntrophin Beta 1. The antigen corresponds to amino acid range 2-297 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20649, 6641
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.