Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNTB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521990 Supplier Invitrogen™ Supplier No. PA521990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Raji. Predicted reactivity: Mouse (96%), Rat (96%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Syntrophins are structurally related proteins located at the neuromuscular synapse and play an important role in dystrophin associated protein (DAP) complex formation. To date there have been three distinct syntrophin isoforms discovered: alpha 1, beta 1, and beta 2. These proteins each contain a PDZ domain, a syntrophin-unique domain (SU), and three pleckstrin homology domains (PH1a,1b and 2). The PDZ domain has been shown to be responsible for binding syntrophin to nNOS and the DAP complex. This association results in localization of the nicotinic acetylcholine receptor to the cytoskeleton via the (S/T)XV C terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNTB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SNTB2
Gene Accession No. Q13425
Gene Alias 59 kDa dystrophin-associated protein A1 basic component 2; beta-2-syntrophin; D16S2531E; dystrophin-associated protein A1, 59kD, basic component 2; EST25263; Snt2; SNT2B2; SNT3; SNTB2; SNTL; SYN; syntrophin beta 2; syntrophin, basic 2; syntrophin, beta 2; syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2); syntrophin-3; Syntrophin-like
Gene Symbols SNTB2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SNTB2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.