Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNTF (cleaved spectrin) Rabbit anti-Mouse, Porcine, Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN226410UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2264-10UL 10 μL
ABN2264-50UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN2264-10UL Supplier MilliporeSigma Supplier No. ABN226410UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SNTF (cleaved spectrin), Cat. No. ABN2264, is a highly specific rabbit polyclonal antibody that targets cleaved Spectrin alpha chain, non-erythrocytic 1 and has been tested for use in ELISA, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNTF (cleaved spectrin)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Spectrin alpha chain non-erythrocytic 1;Alpha-II spectrin;Fodrin alpha chain;Spectrin non-erythroid alpha subunit;Calpain-derived alpha-II spectrin N-terminal fragment
Gene Symbols SPTAN1;NEAS;SPTA2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to calpain-generated neoepitope in SNTF (CAQQEVY).
Molecular Weight of Antigen ∽160 kDa observed. Calculated molecular weight of full length protein is 284.54 kDa. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 10 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_003118
Test Specificity This rabbit polyclonal antibody specifically detects calpain-generated neoepitope in SNTF. It targets a neoepitope of 6 amino acids at the carboxyl-end of the stable calpain-derived alpha-spectrin.
Target Species Mouse, Pig, Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.