Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNTG1 Antibody (OTI3G10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274261F
Description
SNTG1 Monoclonal specifically detects SNTG1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SNTG1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SNTG1 | |
Full length recombinant protein of human SNTG1 (NP_061840) produced in HEK293T cells. | |
0.1 mL | |
54212.0 | |
Store at 4°C in the dark. |
Immunocytochemistry, Immunofluorescence | |
OTI3G10 | |
Immunocytochemistry/Immunofluorescence | |
G1SYNsyntrophin-4, gamma-1-syntrophin, gamma1-syntrophin, SYN4syntrophin 4, syntrophin, gamma 1, Syntrophin-4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction