Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNTG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570303
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
Adapter protein that binds to and probably organizes the subcellular localization of a variety of proteins. May link various receptors to the actin cytoskeleton and the dystrophin glycoprotein complex. May participate in regulating the subcellular location of diacylglycerol kinase-zeta to ensure that diacylglycerol is rapidly inactivated following receptor activation.
Specifications
SNTG1 | |
Polyclonal | |
Unconjugated | |
SNTG1 | |
4933426D16Rik; G1SYN; Gamma-1-syntrophin; gamma1-syntrophin; RGD1560290; Sntg1; SYN4; syntrophin 4; syntrophin gamma 1; syntrophin, gamma 1; syntrophin-4; zmp:0000001117 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NSN8 | |
SNTG1 | |
synthetic peptide directed towards the middle region of human SNTG1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction