Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNTG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567236 Supplier Invitrogen™ Supplier No. PA567236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLHSRFSQYVP GTDLSRQNAF QVIAVDGVCT GIIQCLSAED CVDWLQAIAT NISNLTKHNI KKINRNFPVN QQIVY Highest antigen sequence identity to the following orthologs - mouse 92%, rat 93%.

Adapter protein that binds to and probably organizes the subcellular localization of a variety of proteins. May link various receptors to the actin cytoskeleton and the dystrophin glycoprotein complex. May participate in regulating the subcellular location of diacylglycerol kinase-zeta to ensure that diacylglycerol is rapidly inactivated following receptor activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNTG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNTG1
Gene Accession No. Q9NSN8
Gene Alias 4933426D16Rik; G1SYN; Gamma-1-syntrophin; gamma1-syntrophin; RGD1560290; Sntg1; SYN4; syntrophin 4; syntrophin gamma 1; syntrophin, gamma 1; syntrophin-4; zmp:0000001117
Gene Symbols SNTG1
Host Species Rabbit
Immunogen Recombinant Human SNTG1. Recombinant protein control fragment (Product #RP-107870).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54212
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.