Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNTG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578505
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIPA578505 Supplier Invitrogen™ missing translation for 'supplierNo' PA578505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A549, H1299 Predicted Reactivity: Mouse (86%), Rat (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Syntrophin gamma 2 encodes a protein belonging to the syntrophin family. Syntrophins are cytoplasmic peripheral membrane proteins that bind to components of mechanosenstive sodium channels and the extreme carboxy-terminal domain of dystrophin and dystrophin-related proteins. The PDZ domain of this protein product interacts with a protein component of a mechanosensitive sodium channel that affects channel gating. Absence or reduction of this protein product has been associated with Duchenne muscular dystrophy. There is evidence of alternative splicing yet the full-length nature of these variants has not been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNTG2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene SNTG2
Gene Accession No. Q9NY99
Gene Alias 2210008K22Rik; 9530013L23; BB121248; G2SYN; Gamma-2-syntrophin; gamma2-syntrophin; MGC133174; SNTG2; SYN5; syntrophin; syntrophin 5; syntrophin gamma 2; syntrophin, gamma 2; syntrophin-5
Gene Symbols SNTG2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Syntrophin gamma 2. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54221
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.