Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNTN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563565
Description
Immunogen sequence: KAIAKDLLQT QFRNFAEGQE TKPKYREILS ELDEHTENKL DFEDFMILLL SITVMSDLLQ Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.

Specifications
SNTN | |
Polyclonal | |
Unconjugated | |
SNTN | |
Protein S100-A1-like; S100 calcium-binding protein A1-like; S100A1L; S100AL; S100A-like protein; sentan; sentan, cilia apical structure protein; SNTN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132203 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NMZ2 | |
SNTN | |
Recombinant protein corresponding to Human SNTN. Recombinant protein control fragment (Product #RP-103328). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction