Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNUPN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121591
This item is not returnable.
View return policy
Description
SNUPN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
Specifications
SNUPN | |
Polyclonal | |
Unconjugated | |
SNUPN | |
0610031A09Rik; hypothetical protein LOC515588; KPNBL; RNA U transporter 1; RNA, U transporter 1; Rnut1; Snupn; Snupn1; snurportin 1; Snurportin1; snurportin-1; snurportin-1-like protein; SPN1; wu:fc41a03; zgc:77819 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
66069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q80W37 | |
SNUPN | |
Recombinant protein Snurportin 1. The antigen corresponds to amino acid range 1-329 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction