Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNUPN Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | SNUPN |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119424
|
Invitrogen™
PA5119424 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121590
|
Invitrogen™
PA5121590 |
20 μL |
Each for $209.50
|
|
|||||
Description
SNUPN Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.Specifications
SNUPN | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
SNUPN | |
0610031A09Rik; hypothetical protein LOC515588; KPNBL; RNA U transporter 1; RNA, U transporter 1; Rnut1; Snupn; Snupn1; snurportin 1; Snurportin1; snurportin-1; snurportin-1-like protein; SPN1; wu:fc41a03; zgc:77819 | |
SNUPN | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O95149, Q80W37 | |
10073, 66069 | |
Recombinant protein Snurportin 1. The antigen corresponds to amino acid range 1-360 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title