Learn More
SNUPN Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305568AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The nuclear import of the spliceosomal snRNPs U1, U2, U4 and U5, is dependent on the presence of a complex nuclear localization signal. The latter is composed of the 5′-2,2,7-terminal trimethylguanosine (m3G) cap structure of the U snRNA and the Sm core domain. The protein encoded by this gene interacts specifically with m3G-cap and functions as an snRNP-specific nuclear import receptor. Alternatively spliced transcript variants encoding the same protein have been identified for this gene.Specifications
SNUPN | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95149 | |
SNUPN | |
Between 310 and 360 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SNUPN | |
KPNBL, RNUT1, Snurportin1, RNA U transporter 1, RNA, U transporter 1, snurportin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10073 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.